Ethyl-3-formylbenzoate ≥98%

Supplier: Alfa Aesar
Warning

ALFAH50945.MDEA 175 SGD
ALFAH50945.MD ALFAH50945.03
Ethyl-3-formylbenzoate ≥98%
Ethyl 3-formylbenzoate
Formula: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD08064275
CAS Number: 33745-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR