2-(Trimethylsilyloxy)benzaldehyde 95%

Supplier: Thermo Scientific
Warning

ALFAL13462.06EA 0 SGD
ALFAL13462.06
2-(Trimethylsilyloxy)benzaldehyde 95%
2-(Trimethylsilyloxy)benzaldehyde
Formula: C₁₀H₁₄O₂Si
MW: 194,31 g/mol
Boiling Pt: 142 °C (25 mmHg)
Density: 1,013
Storage Temperature: Ambient
MDL Number: MFCD00040721
CAS Number: 1078-31-5
EINECS: 214-081-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR