5-Chloro-2-(trifluoromethoxy)benzonitrile 97%, JRD

Supplier: Alfa Aesar
Danger

ALFAH31977.03EA 446 SGD
ALFAH31977.03 ALFAH31977.MD
5-Chloro-2-(trifluoromethoxy)benzonitrile 97%, JRD
5-Chloro-2-(trifluoromethoxy)benzonitrile
Formula: C₈H₃ClF₃NO
Melting Pt: lorem ipsum
Storage Temperature: Ambient
MDL Number: MFCD11519364
CAS Number: 1092461-25-0
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR