3-Chloro-4-(trifluoromethoxy)benzonitrile ≥97%

Supplier: Alfa Aesar
Danger

ALFAB23362.06EA 543 SGD
ALFAB23362.06 ALFAB23362.03
3-Chloro-4-(trifluoromethoxy)benzonitrile ≥97%
3-Chloro-4-(trifluoromethoxy)benzonitrile
Formula: C₈H₃ClF₃NO
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD01631557
CAS Number: 129604-26-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR