2-Hydroxy-3-methylbenzhydrazide ≥98%

Supplier: Alfa Aesar
Warning

ALFAL08980.03EA 107 SGD InStock
ALFAL08980.03 ALFAL08980.06
2-Hydroxy-3-methylbenzhydrazide ≥98%
2-Hydroxy-3-methylbenzhydrazide
Formula: C₈H₁₀N₂O₂
MW: 166,18
Melting Pt: 189…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00017052
CAS Number: 30991-42-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR