2-(p-Toluenesulphonyl)ethanol ≥98%
Supplier: Thermo Scientific
Synonyms:
2-Tosylethanol
ALFAA18249.06EA
0
SGD
ALFAA18249.06
2-(p-Toluenesulphonyl)ethanol ≥98%
2-(p-Toluenesulphonyl)ethanol
Formula:
C₉H₁₂O₃S MW: 200,2600 g/mol Melting Pt: 50…53 °C |
MDL Number:
MFCD00041757 CAS Number: 22381-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...