Benzaldehyde azine ≥99%

Supplier: Alfa Aesar
Warning

ALFAA15721.36EA 521 SGD
ALFAA15721.36 ALFAA15721.22 ALFAA15721.14
Benzaldehyde azine ≥99%
Benzaldehyde azine
Formula: C₁₄H₁₂N₂
Melting Pt: 91…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00016876
CAS Number: 588-68-1
EINECS: 209-627-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR