2-Chloro-4,6-bis(trifluoromethyl)benzoic acid 97%
|
Warning
|
ALFAH31784.MDEA
145
SGD
ALFAH31784.MD
ALFAH31784.03
2-Chloro-4,6-bis(trifluoromethyl)benzoic acid 97%
2-Chloro-4,6-bis(trifluoromethyl)benzoic acid
Formula:
C₉H₃ClF₆O₂ MW: 292,56 g/mol Melting Pt: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD09025365 CAS Number: 916420-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...