Potassium-3-(4-morpholinylcarbonyl)phenyltrifluoroborate

Supplier: Alfa Aesar
Warning

ALFAH51142.03EA 107 SGD InStock
ALFAH51142.03 ALFAH51142.06
Potassium-3-(4-morpholinylcarbonyl)phenyltrifluoroborate
Potassium 3-(4-morpholinylcarbonyl)phenyltrifluoroborate
Formula: C₁₁H₁₂BF₃KNO₂
MW: 297,12
Storage Temperature: Ambient
MDL Number: MFCD11977713
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR