Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium-4-(benzylaminocarbonyl)phenyltrifluoroborate ≥95%

Supplier: Alfa Aesar
Warning

ALFAH51887.06EA 239 SGD InStock
ALFAH51887.06 ALFAH51887.03
Potassium-4-(benzylaminocarbonyl)phenyltrifluoroborate ≥95%
Potassium 4-(benzylaminocarbonyl)phenyltrifluoroborate


Formula: C₁₄H₁₂BF₃KNO
MW: 317,16
Storage Temperature: Ambient
MDL Number: MFCD16293900
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty