Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Benzyl-4-methoxyaniline ≥99%

Supplier: Alfa Aesar
Danger

ALFAL11158.03EA 95.6 SGD InStock
ALFAL11158.03 ALFAL11158.06
N-Benzyl-4-methoxyaniline ≥99%
N-Benzyl-4-methoxyaniline


Formula: C₁₄H₁₅NO
MW: 213,28
Boiling Pt: 138…140 °C (0,1 mmHg)
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00059298
CAS Number: 17377-95-6
EINECS: 000-000-0
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty