Dimagnesium silicide ≥99.99% (metals basis)

Supplier: Alfa Aesar
Danger

Synonyms: Magnesium silicide

ALFA45518.30EA 1490 SGD
ALFA45518.30 ALFA45518.18 ALFA45518.09
Dimagnesium silicide ≥99.99% (metals basis)
Dimagnesium silicide
Formula: Mg₂Si
MW: 76,70 g/mol
Melting Pt: 1102 °C
Density: 2,0
Storage Temperature: Ambient
MDL Number: MFCD00016202
CAS Number: 22831-39-6
EINECS: 245-254-5
UN: 2624
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR