Titanium disilicide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), thickness 3.18 mm (0.125 in)

Supplier: Alfa Aesar

Synonyms: Titanium silicide

ALFA41116.KSEA 1840 SGD InStock
ALFA41116.KS
Titanium disilicide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), thickness 3.18 mm (0.125 in)
Titanium disilicide
Formula: Si₂Ti
MW: 104,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151541
CAS Number: 12039-83-7
EINECS: 234-904-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR