Molybdenum disilicide ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Synonyms:
Molybdenum silicide
ALFA41076.KSEA
806
SGD
ALFA41076.KS
Molybdenum disilicide ≥99.5% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Molybdenum disilicide
Magnetron sputtering source
Formula:
MoSi₂ MW: 152,11 g/mol Melting Pt: 1870…2030 °C Density: 6,31 Storage Temperature: Ambient |
MDL Number:
MFCD00151535 CAS Number: 12136-78-6 EINECS: 234-786-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...