Molybdenum disilicide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), thickness 6.35 mm (0.250 in)

Supplier: Alfa Aesar

Synonyms: Molybdenum silicide

ALFA41079.KSEA 1330 SGD InStock
ALFA41079.KS
Molybdenum disilicide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), thickness 6.35 mm (0.250 in)
Molybdenum disilicide
Formula: MoSi₂
MW: 152,11 g/mol
Melting Pt: 1870…2030 °C
Density: 6,31
Storage Temperature: Ambient
MDL Number: MFCD00151535
CAS Number: 12136-78-6
EINECS: 234-786-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR