3-Bromo-2-thiophenecarbonitrile ≥97%

Supplier: Alfa Aesar
Danger

ALFAL09328.MDEA 141 SGD
ALFAL09328.MD ALFAL09328.03
3-Bromo-2-thiophenecarbonitrile ≥97%
3-Bromo-2-thiophenecarbonitrile
Formula: C₅H₂BrNS
Melting Pt: 46…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00068173
CAS Number: 18791-98-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR