Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitro-4-thiophenecarbonitrile ≥97%

Danger

ALFAL11428.03EA 298 SGD InStock
ALFAL11428.03 ALFAL11428.MD
2-Nitro-4-thiophenecarbonitrile ≥97%
2-Nitro-4-thiophenecarbonitrile


Formula: C₅H₂N₂O₂S
MW: 154,15
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00052580
CAS Number: 42137-23-5
EINECS: 000-000-0
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty