2-Nitro-4-thiophenecarboxylic acid ≥98%

Supplier: Alfa Aesar
Warning

ALFAA10705.03EA 938 SGD
ALFAA10705.03 ALFAA10705.MD ALFAA10705.06
2-Nitro-4-thiophenecarboxylic acid ≥98%
2-Nitro-4-thiophenecarboxylic acid
Formula: C₅H₃NO₄S
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00052582
CAS Number: 40357-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR