N-Fmoc-glycine

Supplier: Panreac Química SLU

Synonyms: Fmoc-Gly-OH

PANR15B108.1604EA 188 SGD
PANR15B108.1604 PANR15B108.1603
N-Fmoc-glycine
N-Fmoc-glycine
Formula: C₁₇H₁₅NO₄
MW: 297,3 g/mol
MDL Number: MFCD00037140
CAS Number: 29022-11-5
EINECS: 249-373-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR