2,1,3-Benzothiadiazole-4-sulphonyl chloride ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Benzo[c][1,2,5]thiadiazole-4-sulphonyl chloride
ALFAH50377.03EA
362
SGD
ALFAH50377.03
ALFAH50377.06
2,1,3-Benzothiadiazole-4-sulphonyl chloride ≥97%
2,1,3-Benzothiadiazole-4-sulfonyl chloride
Formula:
C₆H₃ClN₂O₂S₂ MW: 234,68 g/mol Melting Pt: 145…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068049 CAS Number: 73713-79-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...