Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-(1,2,4-Triazol-4-yl)acetanilide ≥99%

Supplier: Alfa Aesar
Warning

ALFAL13628.03EA 68.5 SGD InStock
ALFAL13628.03 ALFAL13628.06
4'-(1,2,4-Triazol-4-yl)acetanilide ≥99%
4'-(1,2,4-Triazol-4-yl)acetanilide


Formula: C₁₀H₁₀N₄O
MW: 202,22
Melting Pt: 284…286 °C
Storage Temperature: Ambient
MDL Number: MFCD01075653
CAS Number: 154594-15-7
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty