4-Isopropylphenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Isopropylbenzeneboronic acid
ALFAL17459.03EA
212
SGD
ALFAL17459.03
ALFAL17459.06
4-Isopropylphenylboronic acid ≥98%
4-Isopropylphenylboronic acid
Formula:
C₉H₁₃BO₂ MW: 164,01 g/mol Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074614 CAS Number: 16152-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...