Chlorobis(4-chlorophenyl)phosphine ≥97%

Supplier: Thermo Scientific
Danger

ALFAH25886.06EA 0 SGD
ALFAH25886.06 ALFAH25886.03
Chlorobis(4-chlorophenyl)phosphine ≥97%
Chlorobis(4-chlorophenyl)phosphine
Formula: C₁₂H₈Cl₃P
MW: 289,53 g/mol
Boiling Pt: 184…186 °C (2,5 mmHg)
Melting Pt: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD09909564
CAS Number: 13685-26-2
EINECS: 000-000-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR