6-Chloro-2,3-difluoroanisole ≥97%

Supplier: Alfa Aesar
Warning

ALFAH34357.06EA 1120 SGD
ALFAH34357.06 ALFAH34357.03
6-Chloro-2,3-difluoroanisole ≥97%
6-Chloro-2,3-difluoroanisole
Formula: C₇H₅ClF₂O
Storage Temperature: Ambient
CAS Number: 1373921-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR