Dibenzyl phthalate ≥97%

Supplier: Alfa Aesar

ALFAL04985.06EA 155 SGD
ALFAL04985.06 ALFAL04985.14
Dibenzyl phthalate ≥97%
Dibenzyl phthalate
Formula: C₂₂H₁₈O₄
Boiling Pt: 276…278 °C (15 mmHg)
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00014437
CAS Number: 523-31-9
EINECS: 208-344-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR