2-Fluoro-4-(trifluoromethoxy)benzonitrile ≥97%
|
Danger
|
ALFAH33876.03EA
109
SGD
ALFAH33876.03
ALFAH33876.06
2-Fluoro-4-(trifluoromethoxy)benzonitrile ≥97%
2-Fluoro-4-(trifluoromethoxy)benzonitrile
Formula:
C₈H₃F₄NO Storage Temperature: Ambient |
MDL Number:
MFCD16652437 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...