5-Fluoro-2-(trifluoromethoxy)benzonitrile 97%, JRD

Supplier: Alfa Aesar
Danger

ALFAH32216.06EA 902 SGD
ALFAH32216.06 ALFAH32216.03
5-Fluoro-2-(trifluoromethoxy)benzonitrile 97%, JRD
5-Fluoro-2-(trifluoromethoxy)benzonitrile
Formula: C₈H₃F₄NO
Storage Temperature: Ambient
MDL Number: MFCD11519332
CAS Number: 1092460-82-6
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR