3-(2-Fluoro-4-(trifluoromethoxy)phenyl)propionic acid ≥97%

Supplier: Alfa Aesar
Warning

Synonyms: Benzenepropanoic acid, 2-fluoro-4-(trifluoromethoxy)-3-[2-Fluoro-4-(trifluoromethoxy)phenyl]propanoic acid

ALFAH34201.MDEA 83.9 SGD InStock
ALFAH34201.MD ALFAH34201.03
3-(2-Fluoro-4-(trifluoromethoxy)phenyl)propionic acid ≥97%
3-(2-Fluoro-4-(trifluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₈F₄O₃
MW: 252,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652497
CAS Number: 1240257-16-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR