3-(4-Methoxy-3-(trifluoromethyl)phenyl)propionic acid 97%, JRD

Supplier: Alfa Aesar
Warning

ALFAH32428.03EA 446 SGD
ALFAH32428.03 ALFAH32428.MD
3-(4-Methoxy-3-(trifluoromethyl)phenyl)propionic acid 97%, JRD
3-(4-Methoxy-3-(trifluoromethyl)phenyl)propionic acid
Formula: C₁₁H₁₁F₃O₃
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD11519317
CAS Number: 1005205-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR