2'-Methyl-4'-nitroacetanilide ≥97%
Supplier: Thermo Scientific
|
Warning
|
ALFAB22054.06EA
0
SGD
ALFAB22054.06
2'-Methyl-4'-nitroacetanilide ≥97%
2'-Methyl-4'-nitroacetanilide
Formula:
C₉H₁₀N₂O₃ MW: 194,19 g/mol Melting Pt: 198…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00033896 CAS Number: 2719-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...