2'-Hydroxy-5'-methylacetanilide ≥97%

Supplier: Alfa Aesar
Warning

ALFAB20630.14EA 286 SGD
ALFAB20630.14 ALFAB20630.06
2'-Hydroxy-5'-methylacetanilide ≥97%
2'-Hydroxy-5'-methylacetanilide
Formula: C₉H₁₁NO₂
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00020183
CAS Number: 6375-17-3
EINECS: 228-934-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR