2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid ≥97%
|
Warning
|
ALFAH33361.03EA
106
SGD
ALFAH33361.03
ALFAH33361.06
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid
Formula:
C₉H₆F₄O₃ MW: 238,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201034 CAS Number: 1373920-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...