2-Methyl-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: Alfa Aesar
Warning

ALFAH26518.06EA 703 SGD
ALFAH26518.06 ALFAH26518.03
2-Methyl-3-(trifluoromethyl)benzoic acid ≥97%
2-Methyl-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
Melting Pt: 106…109 °C
Storage Temperature: Ambient
MDL Number: MFCD01631588
CAS Number: 62089-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR