3-Methyl-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: Alfa Aesar
Warning

ALFAH34410.06EA 756 SGD
ALFAH34410.06 ALFAH34410.03
3-Methyl-5-(trifluoromethyl)benzoic acid ≥97%
3-Methyl-5-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD01631590
CAS Number: 117186-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR