4-Methyl-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: Alfa Aesar
Warning

ALFAB24246.06EA 434 SGD InStock
ALFAB24246.06 ALFAB24246.03
4-Methyl-3-(trifluoromethyl)benzoic acid ≥97%
4-Methyl-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
MW: 204,15
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD01631592
CAS Number: 261952-01-6
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR