Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dibromoisonicotinic acid ≥97%

Supplier: Alfa Aesar
Warning
Synonyms: 2,3-Dibromopyridine-4-carboxylic acid

ALFAH31708.MDEA 233 SGD InStock
ALFAH31708.MD ALFAH31708.03
2,3-Dibromoisonicotinic acid ≥97%
2,3-Dibromoisonicotinic acid


Formula: C₆H₃Br₂NO₂
MW: 280,90
Storage Temperature: Ambient
MDL Number: MFCD09997708
CAS Number: 1020056-98-7
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty