Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Difluoropicolinic acid 97%

Supplier: Alfa Aesar
Warning
Synonyms: 3,5-Difluoropyridine-2-carboxylic acid

ALFAL20324.03EA 154 SGD InStock
ALFAL20324.03 ALFAL20324.06
3,5-Difluoropicolinic acid 97%
3,5-Difluoropicolinic acid


Formula: C₆H₃F₂NO₂
MW: 159,09
Melting Pt: ∼ 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD05664124
CAS Number: 745784-04-7
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty