Dimethyl-4-nitrophthalate ≥98%

Supplier: Alfa Aesar

ALFAA17885.03EA 41.5 SGD
ALFAA17885.03 ALFAA17885.06
Dimethyl-4-nitrophthalate ≥98%
Dimethyl 4-nitrophthalate
Formula: C₁₀H₉NO₆
Boiling Pt: 192…195 °C (25 mmHg)
Melting Pt: 65…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00017191
CAS Number: 610-22-0
EINECS: 210-212-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR