Isopropyldiphenylphosphine ≥99%
Supplier: Thermo Scientific
|
Warning
|
ALFAH27565.06EA
569
SGD
ALFAH27565.06
ALFAH27565.03
Isopropyldiphenylphosphine ≥99%
Isopropyldiphenylphosphine
Formula:
C₁₅H₁₇P MW: 228,27 g/mol Boiling Pt: 165 °C (11 mmHg) Melting Pt: 38…41 °C Density: 0,861 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015028 CAS Number: 6372-40-3 EINECS: 228-902-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...