4-(4-Fluorophenoxy)benzonitrile ≥98%
|
Warning
|
ALFAH32530.22EA
392
SGD
ALFAH32530.22
4-(4-Fluorophenoxy)benzonitrile ≥98%
4-(4-Fluorophenoxy)benzonitrile
Formula:
C₁₃H₈FNO MW: 213,21 g/mol Melting Pt: 67…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD01815090 CAS Number: 215589-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...