2-Methoxyethyl chloroformate tech. 85%, Technical Grade
Supplier: Thermo Scientific
|
Danger
|
ALFAH25937.AAEA
119
SGD
ALFAH25937.AA
ALFAH25937.AC
2-Methoxyethyl chloroformate tech. 85%, Technical Grade
2-Methoxyethyl chloroformate
Formula:
C₄H₇ClO₃ MW: 138,55 g/mol Boiling Pt: 157…158 °C Density: 1,192 Flash Pt: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00058932 CAS Number: 628-12-6 EINECS: 211-026-9 UN: 2742 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...