Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-2-fluoro-3-methylanisole ≥97%

Supplier: Alfa Aesar
Warning

ALFAH26579.06EA 415 SGD InStock
ALFAH26579.06 ALFAH26579.03
6-Chloro-2-fluoro-3-methylanisole ≥97%
6-Chloro-2-fluoro-3-methylanisole


Formula: C₈H₈ClFO
MW: 174,60
Boiling Pt: 66 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01631377
CAS Number: 261762-80-5
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty