Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dicyclohexylphenylphosphine ≥98%

Supplier: Alfa Aesar
Warning

ALFAH26992.03EA 213 SGD InStock
ALFAH26992.03 ALFAH26992.06
Dicyclohexylphenylphosphine ≥98%
Dicyclohexylphenylphosphine


Formula: C₁₈H₂₇P
MW: 274,38
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00003854
CAS Number: 6476-37-5
EINECS: 229-334-7
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty