Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: Alfa Aesar
Warning

ALFAH26339.09EA 846 SGD InStock
ALFAH26339.09 ALFAH26339.03
2-Fluoro-5-(trifluoromethoxy)benzoic acid ≥97%
2-Fluoro-5-(trifluoromethoxy)benzoic acid


Formula: C₈H₄F₄O₃
MW: 224,11
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD04115887
CAS Number: 886497-85-4
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty