Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: Alfa Aesar
Warning

ALFAH33011.06EA 903 SGD InStock
ALFAH33011.06 ALFAH33011.03
3-Fluoro-5-(trifluoromethoxy)benzoic acid ≥97%
3-Fluoro-5-(trifluoromethoxy)benzoic acid


Formula: C₈H₄F₄O₃
MW: 224,11
Storage Temperature: Ambient
MDL Number: MFCD18783221
CAS Number: 1242258-49-6
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty