Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Aspartic acid

Supplier: Panreac Química SLU


PANR202422.0914EA 0 SGD InStock
PANR202422.0914
(±)-Aspartic acid
(±)-Aspartic acid


Formula: C₄H₇NO₄
MW: 133,10
MDL Number: MFCD00063083
CAS Number: 617-45-8
EINECS: 210-513-3
Order Now


 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty