Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-4-fluorosalicylate ≥98%

Supplier: Alfa Aesar
Warning

ALFAL17846.03EA 397 SGD InStock
ALFAL17846.03 ALFAL17846.MD
Methyl-4-fluorosalicylate ≥98%
Methyl 4-fluorosalicylate


Formula: C₈H₇FO₃
MW: 170,14
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD02093965
CAS Number: 392-04-1
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty