Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Butyl-5-ethylthiophene ≥97%

Supplier: Alfa Aesar

ALFAB22708.06EA 100 SGD InStock
ALFAB22708.06 ALFAB22708.14
2-Butyl-5-ethylthiophene ≥97%
2-Butyl-5-ethylthiophene


Formula: C₁₀H₁₆S
MW: 168,30
Storage Temperature: Ambient
MDL Number: MFCD00053118
CAS Number: 54411-06-2
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty