Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butylphenylhydrazine hydrochloride ≥97%

Supplier: Alfa Aesar

ALFAA14385.06EA 374 SGD InStock
ALFAA14385.06
4-tert-Butylphenylhydrazine hydrochloride ≥97%
4-tert-Butylphenylhydrazine hydrochloride


Formula: C₁₀H₁₇ClN₂
Melting Pt: ∼228 °C (dec)
MDL Number: MFCD00013387
CAS Number: 128231-55-0
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty