Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile 97%, JRD

Supplier: Alfa Aesar
Danger

ALFAH31652.03EA 419 SGD InStock
ALFAH31652.03 ALFAH31652.MD
2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile 97%, JRD
2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile


Formula: C₁₀H₅F₆NO
MW: 269,14
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD11519321
CAS Number: 1092460-75-7
EINECS: 000-000-0
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty